Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2α Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS123P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS123P 1 mL Concentrated 200μg/mL
MS123R7 7 mL Prediluted Prediluted
MS123RQ 12 mL Ready-to-Use Prediluted
MS123P0 100 μL Concentrated 200μg/mL
MS123P1ABX 100 μL Concentrated 1mg/mL
MS123PABX 200 μL Concentrated 1mg/mL
MS123P1 500 μL Concentrated 200μg/mL
MS123R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS123P Supplier Epredia Supplier No. MS123P
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and flow cytometry experiments.

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse

Clone: 100/D5

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: aa 41-54

Immunogen: A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein

Molecular Weight: 25-26kDa

Positive Control: Raji Cells, Tonsil or follicular lymphomas

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Specifications

Antigen Bcl-2alpha Ab-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 100/D5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.