Learn More
Description
Host Species: Mouse
Clone: 100/D5
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: aa 41-54
Immunogen: A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein
Molecular Weight: 25-26kDa
Positive Control: Raji Cells, Tonsil or follicular lymphomas
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Flow Cytometry
Specifications
Specifications
| Antigen | Bcl-2alpha Ab-1 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 100/D5 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein |
| Quantity | 200 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.