Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Bcl-2α Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS597P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS597P1ABX 100 μL Concentrated 1mg/mL
MS597B0 100 μL Concentrated 200μg/mL
MS597P0 100 μL Concentrated 200μg/mL
MS597PABX 200 μL Concentrated 1mg/mL
MS597B1 500 μL Concentrated 200μg/mL
MS597P1 500 μL Concentrated 200μg/mL
MS597B 1 mL Concentrated 200μg/mL
MS597R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS597P1ABX Supplier Epredia™ Supplier No. MS597P1ABX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse

Clone: 8C8

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with mouse and rat. Others not tested.

Epitope: aa 41-54

Immunogen: A synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human Bcl-2 protein

Molecular Weight: 25-26kDa

Positive Control: Raji or HeLa Cells, Tonsil or follicular lymphomas

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Bcl-2alpha Ab-3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8C8
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A Synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human bcl-2 protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.