Learn More
Epredia™ Lab Vision™ Biotin Ab-2 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: BTN.4
Isotype: IgG1/κ
Species Reactivity: All species.
Immunogen: Biotin conjugated to BSA
Recommended for:
- Western Blotting
- Confocal Microscopy
- Flow Cytometry
- Image Analysis
- In situ Nucleic Acid Hybridization
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Biotin Ab-2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | BTN.4 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Biotin conjugated to BSA |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.