Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Biotin Ab-2 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide

Catalog No. MS1048P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1048P0 100 μL 200μg/mL
MS1048P1 500 μL 200μg/mL
MS1048P 1 mL 200μg/mL
MS1048P1ABX 100 μL 1mg/mL
MS1048PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1048P0 Supplier Epredia™ Supplier No. MS1048P0

Detect both the free and protein-conjugated forms of biotin with Epredia™ Biotin Ab-2, Mouse Monoclonal Antibody.

Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

Host Species: Mouse

Clone: BTN.4

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Biotin conjugated to BSA

Recommended for:

  • Western Blotting
  • Confocal Microscopy
  • Flow Cytometry
  • Image Analysis
  • In situ Nucleic Acid Hybridization

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Biotin Ab-2
Applications Western Blot
Classification Monoclonal
Clone BTN.4
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Biotin conjugated to BSA
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species All species
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.