Learn More
Epredia™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: BRD.3
Isotype: IgG1
Species Reactivity: All species.
Immunogen: Bromodeoxyuridine (BrdU) conjugated to BSA
Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Bromodeoxyuridine (BrdU) Ab-3 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | BRD.3 |
| Concentration | 40μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Bromodeoxyuridine (BrdU) conjugated to BSA |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.