Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS1058B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Conjugate:
Biotin
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MS1058B1 500 μL Biotin
MS1058B0 100 μL Biotin
MS1058B 1 mL Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1058B1 Supplier Epredia™ Supplier No. MS1058B1

Detect S-phase cells with Epredia™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: BRD.3

Isotype: IgG1

Species Reactivity: All species.

Immunogen: Bromodeoxyuridine (BrdU) conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Bromodeoxyuridine (BrdU) Ab-3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BRD.3
Concentration 40μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Bromodeoxyuridine (BrdU) conjugated to BSA
Quantity 500 μL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species All species
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.