Learn More
Epredia™ Lab Vision™ c-Abl Ab-1 Mouse Monoclonal Antibody, biotin labeled with BSA and azide
Shop All Epredia ProductsDescription
The c-Abl proto-oncogene encodes a protein tyrosine kinase that is located in the cytoplasm and nucleus. In chronic myelogenous leukemia and in a subset of acute lymphoblastic leukemias, the c-Abl proto-oncogene undergoes a (9;22) chromosomal translocation producing a novel rearranged chromosome (the Philadelphia chromosome). As the result of the fusion of c-Abl sequences from chromosome 9 to the Bcr gene on chromosome 22. The molecular consequence of this translocation is the generation of a chimeric Bcr/Abl mRNA encoding activated Abl protein tyrosine kinase.
Host Species: Mouse
Clone: 8E9
Isotype: IgG1/γ
Species Reactivity: Human and Mouse. Others not tested.
Epitope: SH2 domain
Immunogen: Recombinant Abl protein
Molecular Weight: 120kDa
Positive Control: HeLa cells or placenta
Cellular Localization: Cytoplasmic and nuclear
Ab-1 reacts with c-Abl p120 and chimeric Bcr/Abl proteins.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | c-Abl Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 8E9 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Recombinant Abl protein |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.