Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ c-Abl Ab-1 Mouse Monoclonal Antibody, biotin labeled with BSA and azide

Catalog No. MS938B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS938B1 500 μL Concentrated
MS938B0 100 μL Concentrated
MS938B 1 mL Concentrated
MS938R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS938B1 Supplier Epredia™ Supplier No. MS938B1

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation procedures.

The c-Abl proto-oncogene encodes a protein tyrosine kinase that is located in the cytoplasm and nucleus. In chronic myelogenous leukemia and in a subset of acute lymphoblastic leukemias, the c-Abl proto-oncogene undergoes a (9;22) chromosomal translocation producing a novel rearranged chromosome (the Philadelphia chromosome). As the result of the fusion of c-Abl sequences from chromosome 9 to the Bcr gene on chromosome 22. The molecular consequence of this translocation is the generation of a chimeric Bcr/Abl mRNA encoding activated Abl protein tyrosine kinase.

Host Species: Mouse

Clone: 8E9

Isotype: IgG1/γ

Species Reactivity: Human and Mouse. Others not tested.

Epitope: SH2 domain

Immunogen: Recombinant Abl protein

Molecular Weight: 120kDa

Positive Control: HeLa cells or placenta

Cellular Localization: Cytoplasmic and nuclear

Ab-1 reacts with c-Abl p120 and chimeric Bcr/Abl proteins.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen c-Abl Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8E9
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Recombinant Abl protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.