Learn More
Description
Host Species: Mouse
Clone: 2B5
Isotype: IgG1
Species Reactivity: Human, Rat, Mouse. Others not tested.
Epitope: aa 1234-1259
Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein
Molecular Weight: 185kDa
Positive Control: T47D or MAD109 cells
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-3/c-erbB-3 Ab-6 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 2B5 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant rat c-erbB-3/HER-3 oncoprotein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.