Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-erbB-3/HER-3 Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS310PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS310PABX 200 μL Unconjugated 1mg/mL
MS310B0 100 μL Biotin 200μg/mL
MS310B1 500 μL Biotin 200μg/mL
MS310B 1 mL Biotin 200μg/mL
MS310P0 100 μL Unconjugated 200μg/mL
MS310P1 500 μL Unconjugated 200μg/mL
MS310P 1 mL Unconjugated 200μg/mL
MS310P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS310PABX Supplier Epredia Supplier No. MS310PABX

Epredia™ c-erbB-3/HER-3 Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2B5

Isotype: IgG1

Species Reactivity: Human, Rat, Mouse. Others not tested.

Epitope: aa 1234-1259

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MAD109 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-3/c-erbB-3 Ab-6
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2B5
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant rat c-erbB-3/HER-3 oncoprotein
Quantity 200 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.