Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-myc Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS139B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS139B0 100 μL Biotin Concentrated 200μg/mL
MS139B1 500 μL Biotin Concentrated 200μg/mL
MS139B 1 mL Biotin Concentrated 200μg/mL
MS139P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS139PABX 200 μL Unconjugated Concentrated 1mg/mL
MS139P1 500 μL Unconjugated Concentrated 200μg/mL
MS139P0 100 μL Unconjugated Concentrated 200μg/mL
MS139R1 1 mL Unconjugated Prediluted Prediluted
MS139R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS139B0 Supplier Epredia Supplier No. MS139B0
Add to Cart
Add to Cart

Epredia™ c-myc Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors.

Host Species: Mouse

Clone: 9E10.3

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.

Epitope: aa 410-419

Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc

Molecular Weight: 64-67 kDa

Positive Control: Raji cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry
  • Electron Microscopy
  • Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen c-myc Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9E10.3
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
Quantity 100 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.