Learn More
Description
Host Species: Mouse
Clone: 9E10.3
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.
Epitope: aa 410-419
Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
Molecular Weight: 64-67 kDa
Positive Control: Raji cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
- Electron Microscopy
- Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | c-myc Ab-2 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 9E10.3 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.