Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Calcitonin Rabbit Monoclonal Antibody

Catalog No. RM9117R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9117R7 7 mL Prediluted
RM9117S0 100 μL Concentrated
RM9117S1 500 μL Concentrated
RM9117S 1 mL Concentrated
RM9117R1 1 mL Prediluted
RM9117RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RM9117R7 Supplier Epredia™ Supplier No. RM9117R7

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Identify a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors using Epredia™ Calcitonin Rabbit Monoclonal Antibody.

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.

Host Species: Rabbit

Clone: SP17

Speices Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Epitope: 1-32aa

Immunogen: Synthetic human calcitonin 1-32 amino acid peptide

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Calcitonin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP17
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic human calcitonin 1-32 amino acid peptide
Quantity 7 mL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Canine, Equine, Human, Mouse, Non-human Primate, Pig, Rat, Sheep
Form Prediluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.