Learn More
Description
Host Species: Rabbit
Clone: SP17
Speices Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.
Epitope: 1-32aa
Immunogen: Synthetic human calcitonin 1-32 amino acid peptide
Positive Control: Thyroid or medullary CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Calcitonin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP17 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic human calcitonin 1-32 amino acid peptide |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Steroids Hormones and Growth Factors |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.