Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Caldesmon, HMW, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1169PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1169PABX 200 μL Concentrated 1mg/mL
MS1169P 1 mL Concentrated 200μg/mL
MS1169P1ABX 100 μL Concentrated 1mg/mL
MS1169P0 100 μL Concentrated 200μg/mL
MS1169P1 500 μL Concentrated 200μg/mL
MS1169R1 1 mL Prediluted
MS1169R7 7 mL Prediluted
MS1169RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS1169PABX Supplier Epredia™ Supplier No. MS1169PABX
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Caldesmon, HMW Ab-1, Mouse Monoclonal Antibody.

Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.

Host Species: Mouse

Clone: h-CALD

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 150kDa (h-caldesmon)

Positive Control: Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Caldesmon, HMW Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone h-CALD
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Crude human uterus extract
Quantity 200 μL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.