Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Calponin-1 Rabbit Monoclonal Antibody

Catalog No. RM2102R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2102R7 7 mL Prediluted
RM2102S0 100 μL Concentrated
RM2102S 1 mL Concentrated
RM2102R1 1 mL Prediluted
RM2102RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM2102R7 Supplier Epredia™ Supplier No. RM2102R7

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ Calponin-1, Rabbit Monoclonal Antibody.

Calponin, a thin filament associated protein, is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.

Host Species: Rabbit

Clone: EP798Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the C-terminus of human Calponin-1

Molecular Weight: 29/34 kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasm Myoepithelial cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Calponin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP798Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from near the C-terminus of human Calponin-1
Quantity 7 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.