Learn More
Description
Host Species: Rabbit
Clone: EP798Y
Species Reactivity: Human. Others not known.
Immunogen: A synthetic peptide derived from near the C-terminus of human Calponin-1
Molecular Weight: 29/34 kDa
Positive Control: Breast CA
Cellular Localization: Cytoplasm Myoepithelial cells
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Calponin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | EP798Y |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from near the C-terminus of human Calponin-1 |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.