Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Calponin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1168RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1168RQ 12 mL Ready-to-Use
MS1168P0 100 μL Concentrated 200μg/mL
MS1168P 1 mL Concentrated 200μg/mL
MS1168P1ABX 100 μL Concentrated 1mg/mL
MS1168PABX 200 μL Concentrated 1mg/mL
MS1168R7 7 mL Prediluted
MS1168R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1168RQ Supplier Epredia™ Supplier No. MS1168RQ

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Calponin Ab-1, Mouse Monoclonal Antibody.

Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. The expression of calponin is restricted to smooth muscle cells and is a marker of the differentiated contractile phenotype of developing smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions.

Host Species: Mouse

Clone: CALP

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not tested.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 34 kDa

Positive Control: Myoepithelial cells in breast ducts

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Calponin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CALP
Concentration
Conjugate Unconjugated
Host Species Mouse
Immunogen Crude human uterus extract
Quantity 12 mL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.