Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calprtectin-Monocyte/Macrophage/L1 protein Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS148R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS148R7 7 mL Prediluted -
MS148P0 100 μL Concentrated 200μg/mL
MS148P1ABX 100 μL Concentrated 1mg/mL
MS148PABX 200 μL Concentrated 1mg/mL
MS148P1 500 μL Concentrated 200μg/mL
MS148P 1 mL Concentrated 200μg/mL
MS148R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS148R7 Supplier Epredia Supplier No. MS148R7

Epredia™ Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Host Species: Mouse

Clone: MAC 387

Isotype: IgG1

Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.

Epitope: Not determined

Immunogen: Affinity purified monocyte membrane preparation

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Macrophage/L1 Protein/Calprotectin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAC 387
Conjugate Unconjugated
Host Species Mouse
Immunogen Affinity purified monocyte membrane preparation
Quantity 7 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Canine, Feline, Guinea Pig, Human, Mouse, Non-human Primate, Pig, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.