Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Caspase 3 (CPP32) Ab-4 Rabbit Polyclonal Antibody, biotin labeled with BSA and azide

Catalog No. RB1197B Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
Ready-to-use
Ready-to-use for IHC (UVLP)
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Formulation
RB1197B 1 mL Biotin Concentrated Purified with BSA and Azide
RB1197B0 100 μL Biotin Concentrated Purified with BSA and Azide
RB1197P0 100 μL Unconjugated Concentrated Purified with BSA and Azide
RB1197B1 500 μL Biotin Concentrated Purified with BSA and Azide
RB1197P1 500 μL Unconjugated Concentrated Purified with BSA and Azide
RB1197P1ABX 500 μL Unconjugated Concentrated Purified w/o BSA and Azide
RB1197P 1 mL Unconjugated Concentrated Purified with BSA and Azide
RB1197PABX 1 mL Unconjugated Concentrated Purified w/o BSA and Azide
RB1197R1 1 mL Unconjugated Prediluted Ready-to-use for IHC (UVLP)
RB1197R7 7 mL Unconjugated Prediluted Ready-to-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. RB1197B Supplier Epredia™ Supplier No. RB1197B

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation procedures.

Caspase 3 is ubiquitously expressed and like other caspases is synthesized as an inactive (32 kDa) proenzyme. Upon activation, Caspase 3 is cleaved at Asp28-Ser29 and Asp175-Ser176 thereby generating two subunits of 17 kDa and 12 kDa, respectively. Recent studies have implicated that Caspase 3 is associated with the induction of apoptosis. Activation of Caspase 3 occurs in response to a variety of apoptotic inducers including Fas-mediated apoptosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Monkey, Rabbit, Hamster, Cow, Sheep, Pig and Dog. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human caspase 3 protein

Molecular Weight: 32 kDa

Positive Control: Jurkat cells, Tonsil

Cellular Localization: Predominantly cytoplasmic with some nuclear staining

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Caspase 3 (CPP32) Ab-4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration 1mg/mL
Conjugate Biotin
Formulation Purified with BSA and Azide
Host Species Rabbit
Immunogen Recombinant full length human caspase 3 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Bovine, Canine, Hamster, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.