Learn More
Epredia™ Lab Vision™ Caspase 3 (CPP32) Ab-4 Rabbit Polyclonal Antibody, biotin labeled with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Monkey, Rabbit, Hamster, Cow, Sheep, Pig and Dog. Others not tested.
Epitope: Not determined
Immunogen: Recombinant full-length human caspase 3 protein
Molecular Weight: 32 kDa
Positive Control: Jurkat cells, Tonsil
Cellular Localization: Predominantly cytoplasmic with some nuclear staining
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Caspase 3 (CPP32) Ab-4 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified w/o BSA and Azide |
| Host Species | Rabbit |
| Immunogen | Recombinant full length human caspase 3 protein |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.