Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Caspase 6 (Mch 2) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and Azide

Catalog No. MS1126P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1126P 1 mL 200μg/mL
MS1126P0 100 μL 200μg/mL
MS1126P1 500 μL 200μg/mL
MS1126P1ABX 100 μL 1mg/mL
MS1126PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1126P Supplier Epredia™ Supplier No. MS1126P

Ensure accurate, reproducible results in western blotting and immunoprecipitation procedures with Epredia™ Caspase 6 (Mch 2) Ab-3, Mouse Monoclonal Antibody.

Caspases play a major role in the transduction of the apoptotic signal and execution of apoptosis in mammalian cells. Caspase 3 (CPP32) and Caspase 6 (Mch2) are the major active caspases in apoptotic cells, and are activated in response to distinct apoptosis-inducing stimuli in all cell lines analyzed. Both CPP32 and Mch2 are present in apoptotic cells as multiple active species. Caspase-6 cleaves nuclear mitotic apparatus protein (NuMA) and mediates the shrinkage and fragmentation of nuclei.

Host Species: Mouse

Clone: 6CSP03; same as 14.1.190

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Caspase 6 prodomain

Immunogen: Recombinant human caspase 6 prodomain

Molecular Weight: 34 kDa

Positive Control: Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Caspase 6 (Mch 2) Ab-3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6CSP03; same as 14.1.190
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human caspase 6 prodomain
Quantity 1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.