Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ β-Catenin Rabbit Monoclonal Antibody

Catalog No. RM2101S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Ready-to-Use
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2101S0 100 μL Concentrated
RM2101S 1 mL Concentrated
RM2101RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RM2101S0 Supplier Epredia™ Supplier No. RM2101S0

Ensure accurate and reproducible results in immunohistochemistry applications with this antibody, part of a comprehensive antibody portfolio.

The catenins (α, β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion. Mutations in the catenin gene, CTNNB1, have been implicated in its accumulation and various forms of carcinomas.

Host Species: Rabbit

Clone: E247

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the N-terminus of human β-catenin

Molecular Weight: 92 kDa

Positive Control: Breast CA

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Catenin, beta
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E247
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from near the N-terminus of human beta-catenin
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.