Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Catenin Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB9035P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9035P1 500 μL Concentrated
RB9035P0 100 μL Concentrated
RB9035P 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RB9035P1 Supplier Epredia Supplier No. RB9035P1

Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Catenin, β, Rabbit Polyclonal Antibody.

The catenins (α, Β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. Β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the β-catenin protein

Molecular Weight: 92kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Cytoplasmic and cell membrane

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Specifications

Antigen Catenin, beta
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide from the C-terminus of the beta-catenin protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.