Learn More
Epredia™ Lab Vision™ CD115/c-fms/CSF-1R/M-CSFR Ab-1 Rat Monoclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rat
Clone: 2-4A5-4
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and cat. Others not known.
Epitope: aa 349-512
Immunogen: v-ras-transformed rat NRK cells expressing transduced human CSF-1R
Molecular Weight: 130 and 150 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Acetone-fixed frozen)
- Immunofluorescence
- Blocks Ligand Binding
- Blocks Ligand-dependent Proliferation
- Inhibits CSF-1-independent Growth of Cells Expressing Mutant CSF-1R
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD115 |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 2-4A5-4 |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Immunogen | v-ras-transformed rat NRK cells expressing transduced human CSF-1R |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.