Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD115/c-fms/CSF-1R/M-CSFR Ab-1 Rat Monoclonal Antibody with BSA and azide

Catalog No. RT601P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Concentration:
1mg/mL
200µg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
RT601P1 500 μL 200µg/mL
RT601P0 100 μL 200µg/mL
RT601P 1 mL 200µg/mL
RT601P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RT601P1 Supplier Epredia™ Supplier No. RT601P1

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

CD115, also known as c-fms, macrophage Colony Stimulating Factor Receptor (M-CSFR), or CSF-1R, exists in immature (gp130c-fms) and mature (gp150c-fms) forms. CSF-1R exhibits an intrinsic tyrosine-specific protein kinase activity. It is expressed on the cells of the mononuclear phagocyte lineage. CSF-1R is an integral transmembrane glycoprotein consisting of a 512 amino acid extracellular domain that includes the CSF-1 binding site, a single 25 amino acid membrane-spanning segment, and a 435 amino acid cytoplasmic portion that includes the tyrosine kinase domain.

Host Species: Rat

Clone: 2-4A5-4

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and cat. Others not known.

Epitope: aa 349-512

Immunogen: v-ras-transformed rat NRK cells expressing transduced human CSF-1R

Molecular Weight: 130 and 150 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen)
  • Immunofluorescence
  • Blocks Ligand Binding
  • Blocks Ligand-dependent Proliferation
  • Inhibits CSF-1-independent Growth of Cells Expressing Mutant CSF-1R

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD115
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2-4A5-4
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen v-ras-transformed rat NRK cells expressing transduced human CSF-1R
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.