Learn More
Description
Host Species: Mouse
Clone: K45
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Extracellular second N-terminus Ig-like domain of c-kit
Immunogen: Recombinant human extracellular c-kit fragment
Molecular Weight: 145kDa
Positive Control: Skin, GI Stromal Tumor (GIST)
Cellular Localization: Membrane/cytoplasmic
Recommended for:
- Immunohistochemistry (Frozen or fixed in methacarn or Stieve's and embedded in paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD117/c-Kit/SCF-Receptor Ab-3 |
| Applications | Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | K45 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human extracellular c-kit fragment |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.