Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117/c-Kit/SCF-Receptor Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS289P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS289P1 500 μL 200μg/mL
MS289PABX 200 μL 1mg/mL
MS289P0 100 μL 200μg/mL
MS289P 1 mL 200μg/mL
MS289P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS289P1 Supplier Epredia Supplier No. MS289P1
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells. Ab-3 does not interfere with the binding of SCF to c-kit

Host Species: Mouse

Clone: K45

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Extracellular second N-terminus Ig-like domain of c-kit

Immunogen: Recombinant human extracellular c-kit fragment

Molecular Weight: 145kDa

Positive Control: Skin, GI Stromal Tumor (GIST)

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen or fixed in methacarn or Stieve's and embedded in paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD117/c-Kit/SCF-Receptor Ab-3
Applications Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone K45
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human extracellular c-kit fragment
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.