Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD117/c-Kit/SCF-Receptor Ab-6 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB1518R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted, Ready-to-Use
Concentration:
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
RB1518R1 1 mL Prediluted, Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB1518R1 Supplier Epredia™ Supplier No. RB1518R1

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody.

c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells Antibody to CD117/c-kit may be helpful in the histopathological identification of gastrointestinal stromal tumors (GIST).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human CD117/c-kit protein

Molecular Weight: 145kDa

Positive Control: Tonsil, GI Stromal Tumor (GIST)

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD117/c-Kit/SCF-Receptor Ab-6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of human CD117/c-kit protein
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Prediluted, Ready-to-Use
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.