Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117/c-Kit/SCF-Receptor Rabbit Polyclonal Antibody, Epredia™

Optimize staining of formalin-fixed, paraffin-embedded tissues with Epredia™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody.

Supplier:  Epredia RB9038P0

 View more versions of this product

Catalog No. RB9038P0


Only null left
Add to Cart

Description

Description

Recognizes a protein of 145kDa, which is identified as CD117/p145kit. This rabbit polyclonal antibody does not interfere with the binding of SCF to c-kit. It precipitates both the unoccupied as well as the occupied form of c-kit. The binding of the stem cell factor (SCF) to the c-kit-encoded receptor tyrosine kinase (Type III) stimulates a variety of biochemical responses that culminate in cellular proliferation, migration, or survival. C-kit plays an important role in hematopoiesis, melanogenesis, and gametogenesis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Dog. Others not tested

Epitope: Cytoplasmic domain

Immunogen: A synthetic peptide from the cytoplasmic domain of human CD117/c-kit protein

Molecular Weight: 145 kDa

Positive Control: Stromal tumor, skin or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD117/c-Kit/SCF-Receptor
Polyclonal
200µg/mL
Rabbit
100 μL
Primary
Concentrated
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
A synthetic peptide from the cytoplasmic domain of human CD117 / c-kit protein
IVD
Canine
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.