Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 (4-1BB) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS621PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS621PABX 200 μL Unconjugated 1mg/mL
MS621B0 100 μL Biotin 200μg/mL
MS621B1 500 μL Biotin 200μg/mL
MS621B 1 mL Biotin 200μg/mL
MS621P0 100 μL Unconjugated 200μg/mL
MS621P1 500 μL Unconjugated 200μg/mL
MS621P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS621PABX Supplier Epredia Supplier No. MS621PABX

Epredia™ CD137 (4-1BB) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

4-1BB (also known as CD137) is an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. It exists as both a monomer and a dimer on the surface of activated T cells. 4-1BB is structurally related to the members of NGFR/TNFR superfamily which are characterized by the presence of three-six patterns of a cysteine-rich motif in their extracellular domains. Other members of this family include low affinity NGFR, two receptors for TNF (TNFR-I & TNFR-II), CD30, CD40, OX40, Fas, and CD27. These molecules are involved in cell growth, survival, and death processes. The cytoplasmic domain of 4-1BB include two runs of acidic amino acids, a potential p56lck binding site, five consecutive glycines at the C-terminus, and four potential phosphorylation sites: one tyrosine, two threonine, and one serine.

Host Species: Mouse

Clone: BBK-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Ectodomain

Immunogen: Ectodomain of human 4-1BB recombinant protein

Molecular Weight: 30kDa

Positive Control: Spleen, thymus or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD137 Ab-1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone BBK-2
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Ectodomain of human 4-1BB recombinant protein
Quantity 200 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.