Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD138 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1793S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1793S0 100 μL Concentrated
MS1793S1 500 μL Concentrated
MS1793S 1 mL Concentrated
MS1793R7 7 mL Prediluted
MS1793RQ 12 mL Ready-to-Use
MS1793R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1793S0 Supplier Epredia™ Supplier No. MS1793S0
Add to Cart
Add to Cart

Obtain accurate and reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence, it is a useful marker for viable myeloma cells.

Host Species: Mouse

Clone: MI15

Isotype: IgG1/κ

Species Reactivity: Human. Others not known

Epitope: Ectodomain

Immunogen: U-266 cells

Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)

Positive Control: U-266 cells, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Specifications

Antigen CD138 Ab-2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MI15
Conjugate Unconjugated
Host Species Mouse
Immunogen U-266 cells
Purification Method U-266 cells, tonsil
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.