Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD141/Thrombomodulin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1102S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1102S 1 mL Concentrated
MS1102S1 500 μL Concentrated
MS1102S0 100 μL Concentrated
MS1102R1 1 mL Prediluted
MS1102R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1102S Supplier Epredia™ Supplier No. MS1102S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Optimize immunohistochemical staining of formalin-fixed tissue using Epredia™ CD141/Thrombomodulin Ab-1, Mouse Monoclonal Antibody.

Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.

Host Species: Mouse

Clone: 141C01; same as 1009

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: EGF domain

Immunogen: Recombinant protein encoding the six repeated EGF domains of human thrombomodulin

Molecular Weight: 75kDa

Positive Control: Mesothelioma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD141
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 141C01; same as 1009
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the six repeated EGF domains of human thrombomodulin
Purification Method Mesothelioma
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.