Learn More
Description
Host Species: Mouse
Clone: 141C01; same as 1009
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: EGF domain
Immunogen: Recombinant protein encoding the six repeated EGF domains of human thrombomodulin
Molecular Weight: 75kDa
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD141 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 141C01; same as 1009 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein encoding the six repeated EGF domains of human thrombomodulin |
| Purification Method | Mesothelioma |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.