Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15 Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1259P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1259P1ABX 100 μL Concentrated 1mg/mL
MS1259P0 100 μL Concentrated 200μg/mL
MS1259P1 500 μL Concentrated 200μg/mL
MS1259P 1 mL Concentrated 200μg/mL
MS1259PABX 200 μL Concentrated 1mg/mL
MS1259R7 7 mL Prediluted
MS1259RQ 12 mL Ready-to-Use
MS1259R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS1259P1ABX Supplier Epredia Supplier No. MS1259P1ABX
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures with Epredia™ CD15 Ab-3, Mouse Monoclonal Antibody.

3-fucosyl-N-acetyllactosamine (3-FAL) or CD15 or X-hapten plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Host Species: Mouse

Clone: MMA; same as LeuM1

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: U937 histiocytic cell line

Molecular Weight: 220kDa

Positive Control: Reed-Sternberg's cells in Hodgkin's lymphoma

Cellular Localization: Membrane and granular paranuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Specifications

Antigen CD15 Ab-3
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MMA; same as LeuM1
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen U937 histiocytic cell line
Quantity 100 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.