Learn More
Description
Host Species: Mouse
Clone: D171
Isotype: IgG1
Species Reactivity: Human and Monkey. Does not react with pig, dog, rabbit, rat, hamster and mouse. Others not known.
Epitope: aa 35-50
Immunogen: HeLa cells
Molecular Weight: 60-80kDa
Positive Control: HeLa, SV 80, HEp-2, RD, A431, HL-60, U-937 cells; Monkey Vero, GMK, or TC-7 cells
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not suitable)
- Immunofluorescence
- Flow Cytometry
- Inhibits Replication of Poliovirus in HeLa Cells
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD155 Ab-1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | D171 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | HeLa cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.