Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD155/PVR (Poliovirus Receptor) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS465PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS465PABX 200 μL Unconjugated 1mg/mL
MS465P1 500 μL Unconjugated 200μg/mL
MS465B0 100 μL Biotin 200μg/mL
MS465B1 500 μL Biotin 200μg/mL
MS465B 1 mL Biotin 200μg/mL
MS465P 1 mL Unconjugated 200μg/mL
MS465P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS465PABX Supplier Epredia Supplier No. MS465PABX
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunofluorescence and flow cytometry applications.

PVR is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from a single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or α and κ,) encode cell surface molecules of about 43kDa and 45kDa polypeptide backbones, respectively. Two other splice variants, Β and γ, encode secreted molecules of about 44kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues.

Host Species: Mouse

Clone: D171

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with pig, dog, rabbit, rat, hamster and mouse. Others not known.

Epitope: aa 35-50

Immunogen: HeLa cells

Molecular Weight: 60-80kDa

Positive Control: HeLa, SV 80, HEp-2, RD, A431, HL-60, U-937 cells; Monkey Vero, GMK, or TC-7 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits Replication of Poliovirus in HeLa Cells

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD155 Ab-1
Applications Immunofluorescence
Classification Monoclonal
Clone D171
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen HeLa cells
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.