Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD163 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1103R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1103R7 7 mL Prediluted
MS1103S0 100 μL Concentrated
MS1103S1 500 μL Concentrated
MS1103S 1 mL Concentrated
MS1103R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1103R7 Supplier Epredia™ Supplier No. MS1103R7
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD163 Ab-1, Mouse Monoclonal Antibody.

CD163 antigen is a type I membrane protein also known as M130 antigen, Ber-Mac3, Ki-M8 or SM4. CD163 is restricted in its expression to the monocytic/macrophage lineage. It is present on all circulating monocytes and most tissue macrophages except those found in the mantle zone and germinal centers of lymphoid follicles, interdigitating reticulum cells and Langerhan's cells.

Host Species: Mouse

Clone: 10D6

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Domains 1 to 4 of the N-terminal region

Immunogen: Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163

Positive Control: Placenta

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD163 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10D6
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163
Quantity 7 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.