Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1a Ab-5 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1856P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1856P 1 mL Concentrated 200μg/mL
MS1856P0 100 μL Concentrated 200μg/mL
MS1856P1 500 μL Concentrated 200μg/mL
MS1856RQ 12 mL Ready-to-Use
MS1856PABX 200 μL Concentrated 1mg/mL
MS1856R1 1 mL Prediluted
MS1856R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1856P Supplier Epredia Supplier No. MS1856P
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry applications with Epredia™ CD1a Ab-5, Mouse Monoclonal Antibody.

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: O10

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Molecular Weight: 49kDa

Positive Control: Tonsil

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD1a Ab-5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone O10
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 1 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.