Learn More
Description
Host Species: Mouse
Clone: O10
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Molecular Weight: 49kDa
Positive Control: Tonsil
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD1a Ab-5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | O10 |
| Concentration | — |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.