Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD23 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS729R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS729R1 1 mL Prediluted
MS729S1 500 μL Concentrated
MS729S 1 mL Concentrated
MS729RQ 12 mL Ready-to-Use
MS729S0 100 μL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS729R1 Supplier Epredia™ Supplier No. MS729R1

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD23 Ab-1, Mouse Monoclonal Antibody.

CD23 is identical to low affinity IgE receptor found on B cells. CD23 is expressed on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. CD23 is also detected in neoplastic cells from cases of B cell chronic lymphocytic leukemia and some cases on centroblastic/centrocytic lymphoma.

Host Species: Mouse

Clone: 1B12

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant external domain of CD23 protein

Molecular Weight: 45 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD23 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1B12
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant external domain of CD23 protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.