Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD29 (Integrin β-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS596P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS596P0 100 μL 200μg/mL
MS596P1 500 μL 200μg/mL
MS596P 1 mL 200μg/mL
MS596P1ABX 100 μL 1mg/mL
MS596PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS596P0 Supplier Epredia Supplier No. MS596P0
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments with Epredia™ CD29 (Integrin β-1) Ab-1.

CD29 (integrin Beta-1/gpIIa) is a cell adhesion molecule appearing on platelets, as the common Beta subunit of the very late activation antigen (VLA), and as a component of various protein complexes binding to extracellular matrix proteins. CD29 is expressed at the cell surface exclusively as part of a heterodimer, in association with one of at least nine different integrin alpha subunits (alpha1, alpha2, alpha3, alpha4, alpha5, alpha6, alpha7, alpha8, and alphav). With the exception of red blood cells and possible weak expression on granulocytes, CD29 is expressed on nearly all cell and tissue types.

Host Species: Mouse

Clone: 4B7R

Isotype: IgG1/κ

Species Reactivity: Human and Pig. Others not tested.

Epitope: Not determined

Immunogen: Human melanoma V+B2 cell line

Molecular Weight: 110kDa (nonreducing) and 130 (reducing)

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD29 (Integrin beta-1) Ab-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 4B7R
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human melanoma VplusB2 cell line
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human, Pig
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.