Learn More
Description
Host Species: Mouse
Clone: 4B7R
Isotype: IgG1/κ
Species Reactivity: Human and Pig. Others not tested.
Epitope: Not determined
Immunogen: Human melanoma V+B2 cell line
Molecular Weight: 110kDa (nonreducing) and 130 (reducing)
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen)
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD29 (Integrin beta-1) Ab-1 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 4B7R |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human melanoma VplusB2 cell line |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.