Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD3 (Early T-Cell Marker) Rabbit Monoclonal Antibody,

Catalog No. RM9107RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9107RQ 12 mL Ready-to-Use
RM9107S0 100 μL Concentrated
RM9107S1 500 μL Concentrated
RM9107S 1 mL Concentrated
RM9107R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9107RQ Supplier Epredia™ Supplier No. RM9107RQ

Stain normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues with Epredia™ CD3 (Early T-Cell Marker), Rabbit Monoclonal Antibody.

This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Clone: SP7

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey, Horse, Dog and Cat. Others not tested.

Epitope: 156-168aa

Immunogen: A synthetic 13-mer peptide corresponding to aa 156-168 of the ε-chain of human CD3 protein

Positive Control: Jurkat cells, Tonsil

Cellular Localization: Membrane

Clone SP7 is excellent for staining of formal/paraffin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP7
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein
Quantity 12 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.