Learn More
Epredia™ Lab Vision™ CD3 (Early T-Cell Marker) Rabbit Monoclonal Antibody,
Shop All Epredia ProductsDescription
Host Species: Rabbit
Clone: SP7
Isotype: IgG
Species Reactivity: Human, Baboon, Monkey, Horse, Dog and Cat. Others not tested.
Epitope: 156-168aa
Immunogen: A synthetic 13-mer peptide corresponding to aa 156-168 of the ε-chain of human CD3 protein
Positive Control: Jurkat cells, Tonsil
Cellular Localization: Membrane
Clone SP7 is excellent for staining of formal/paraffin.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP7 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.