Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD3ε Rabbit Monoclonal Antibody

Catalog No. RM2109RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2109RQ 12 mL Ready-to-Use
RM2109S 1 mL Concentrated
RM2109S0 100 μL Concentrated
RM2109R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM2109RQ Supplier Epredia™ Supplier No. RM2109RQ

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD3ε, Rabbit Monoclonal Antibody.

CD3 consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in the majority of T cell neoplasms.

Host Species: Rabbit

Clone: EP449E

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues in the cytoplasmic domain of human CD3ε. The antibody does not cross react with other subunits of CD3.

Molecular Weight: 23kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD3e
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP449E
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in the cytoplasmic domain of human CD3epsilon. The antibody does not cross react with other subunits of CD3.
Quantity 12 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.