Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1 (Endothelial Cell Marker) Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB10333R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB10333R7 7 mL Prediluted
RB10333P1 500 μL Concentrated
RB10333P 1 mL Concentrated
RB10333R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB10333R7 Supplier Epredia Supplier No. RB10333R7

Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody.

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion. Antibody to CD31 is of value in the study of benign and malignant vascular tumors. Staining for CD31 has also been used to measure angiogenesis, which reportedly predicts tumor recurrence.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminus of mouse CD31 protein

Molecular Weight: 100 and 130 kDa

Positive Control: Tonsil, Placenta

Cellular Localization: Endothelial cells in blood vessel of tonsil

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD31 (Endothelial Cell Marker)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of mouse CD31 protein
Purification Method Tonsil
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.