Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35/CR1 (Follicular Dendritic Cell Marker) Mouse Monoclonal Antibody, Epredia™

Catalog No. MS336P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
800μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS336P 1 mL Concentrated 800μg/mL
MS336P0 100 μL Concentrated 800μg/mL
MS336P1 500 μL Concentrated 800μg/mL
MS336P1ABX 100 μL Concentrated 1mg/mL
MS336R7 7 mL Prediluted
MS336PABX 200 μL Concentrated 1mg/mL
MS336R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS336P Supplier Epredia Supplier No. MS336P

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

Complement receptor 1 (CR1, CD35) is a cell membrane-bound, monomeric glycoprotein that is found on numerous cell types of primates including erythrocytes, lymphocytes, and phagocytes. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules.

Host Species: Mouse

Clone: E11

Isotype: IgG1

Species Reactivity: Human, Rhesus Monkey and Baboon. Others not tested.

Epitope: Distal from the C3b-binding site

Immunogen: Intact human monocytes

Molecular Weight: 210-220 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD35
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E11
Concentration 800μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Intact human monocytes
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.