Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36/GPIIIb/GPIV Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS756P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS756P 1 mL 200μg/mL
MS756P1ABX 100 μL 1mg/mL
MS756P1 500 μL 200μg/mL
MS756PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS756P Supplier Epredia Supplier No. MS756P

Ensure accurate, reproducible results in immunohistochemistry , immunofluorescence, flow cytometry and function blocking experiment.

CD36 (also known as platelet glycoprotein IV, OKM5-antigen, or platelet glycoprotein IIIb) acts as a receptor for thrombospondin (TSP), collagen, falciparum malaria parasitized erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL), and apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischaemia observed in sickle cell disease and cerebral malaria. Ab-3 blocks adhesion of P. falciparum parasitized red blood cells to CD36 and strongly inhibits collagen-induced platelet aggregation.

Host Species: Mouse

Clone: 185-1G2

Isotype: IgG2a/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 88 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Function Blocking

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD36/GPIIIb/GPIV Ab-3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 185-1G2
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Stimulated human leukocytes
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.